missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methylphenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62006.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496916

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methylphenyl)-3,5-difluorobenzenesulfonamide
317.74
MFCD26960671

Specifications

N-(3-Chloro-4-methylphenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD26960671
Solid
317.74
1g
Videos
SDS