missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methylphenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59854.03

Code : NEW

Additional Details : CAS Number : 331445-10-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416288

  • £358.49 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methylphenyl)-2-fluorobenzamide
331445-10-4
263.7
MFCD00808716

Specifications

N-(3-Chloro-4-methylphenyl)-2-fluorobenzamide
Solid
263.7
1g
331445-10-4
97%
MFCD00808716
Videos
SDS