missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methylphenyl)-2,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61090.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476256

  • £68.26 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methylphenyl)-2,5-difluorobenzamide
281.69
MFCD11224023

Specifications

N-(3-Chloro-4-methylphenyl)-2,5-difluorobenzamide
97%
MFCD11224023
Solid
281.69
1g
Videos
SDS