missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methoxyphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59087.03

Code : NEW

Additional Details : CAS Number : 413582-01-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15464766

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methoxyphenyl)-3-methylbenzamide
413582-01-1
275.73
MFCD00425024

Specifications

N-(3-Chloro-4-methoxyphenyl)-3-methylbenzamide
Solid
275.73
1g
413582-01-1
97%
MFCD00425024
Videos
SDS