Alfa Aesar™ N-(3-Chloro-4-methoxyphenyl)-2-methoxybenzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59017.MD

250MG N-(3-Chloro-4-methoxyphenyl)-2-methoxybenzamide, 97% 250mg

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 331446-79-8 Weight: 0.00025kg


Quantity 250mg

Product Code. 15404726

Quantity Price
1 £ 111.72 / 250mg
Estimated Shipment date
from Supplier 31-01-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(3-Chloro-4-methoxyphenyl)-2-methoxybenzamide
CAS 331446-79-8
Formula Weight 291.73
MDL Number MFCD01076856

Specification

Formula Weight 291.73
Chemical Name or Material N-(3-Chloro-4-methoxyphenyl)-2-methoxybenzamide
Physical Form Solid
Assay 97%
MDL Number MFCD01076856
CAS 331446-79-8
Quantity 250mg