missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60090.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496558

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-fluorophenyl)benzenesulfonamide
285.72
MFCD01213709

Specifications

N-(3-Chloro-4-fluorophenyl)benzenesulfonamide
97%
MFCD01213709
Solid
285.72
1g
Videos
SDS