missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-fluorophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61909.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456846

  • £63.71 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-fluorophenyl)-3,5-difluorobenzamide
285.65
MFCD05474564

Specifications

N-(3-Chloro-4-fluorophenyl)-3,5-difluorobenzamide
97%
MFCD05474564
Solid
285.65
1g
Videos
SDS