missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-fluorophenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61606.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416646

  • £68.26 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-fluorophenyl)-3,4-difluorobenzenesulfonamide
321.7
MFCD05116037

Specifications

N-(3-Chloro-4-fluorophenyl)-3,4-difluorobenzenesulfonamide
97%
MFCD05116037
Solid
321.7
1g
Videos
SDS