missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-fluorophenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59038.03

Code : NEW

Additional Details : CAS Number : 349128-65-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445218

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-fluorophenyl)-2-fluorobenzamide
349128-65-0
267.66
MFCD01214109

Specifications

N-(3-Chloro-4-fluorophenyl)-2-fluorobenzamide
Solid
267.66
1g
349128-65-0
97%
MFCD01214109
Videos
SDS