missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-fluorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65329.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439566

  • £85.44 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-fluorophenyl)-2,2-diphenylacetamide
339.79
MFCD01269147

Specifications

N-(3-Chloro-4-fluorophenyl)-2,2-diphenylacetamide
95%
MFCD01269147
Solid
339.79
1g
Videos
SDS