missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-2-methylphenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59219.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15404896

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-2-methylphenyl)-3,5-dinitrobenzamide
335.7
MFCD00590777

Specifications

N-(3-Chloro-2-methylphenyl)-3,5-dinitrobenzamide
97%
MFCD00590777
Solid
335.7
1g
Videos
SDS