missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Bromophenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65544.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439726

  • £85.44 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Bromophenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide
407.64
MFCD28098804

Specifications

N-(3-Bromophenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide
Solid
407.64
1g
95%
MFCD28098804
Videos
SDS