missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Bromophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61644.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418208

  • £68.26 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Bromophenyl)-3,5-difluorobenzamide
312.11
MFCD05474336

Specifications

N-(3-Bromophenyl)-3,5-difluorobenzamide
Solid
312.11
1g
97%
MFCD05474336
Videos
SDS