missing translation for 'onlineSavingsMsg'
Learn More

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, Thermo Scientific Chemicals

Catalog Number p-3776795
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7663-77-6
C7H14N2O
142.20
MFCD00003201
HJORCZCMNWLHMB-UHFFFAOYSA-N
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
82111
1-(3-aminopropyl)pyrrolidin-2-one
NCCCN1CCCC1=O
This item is not returnable. View return policy
7663-77-6
C7H14N2O
142.20
MFCD00003201
HJORCZCMNWLHMB-UHFFFAOYSA-N
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
82111
1-(3-aminopropyl)pyrrolidin-2-one
NCCCN1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.0100g/mL
Boiling Point 120.0°C to 123.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 1,5-Diazabicyclo(4.3.0.)non-5-ene: Balance (GC)
Packaging Glass bottle
Refractive Index 1.4970 to 1.4990
Quantity 5 g
Specific Gravity 1.01
Formula Weight 142.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-(3'-Aminopropyl)-2-pyrrolidinone
Show More Show Less
compliance-icons
Product Identifier
  • N-(3'-Aminopropyl)-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'