missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,5-Difluorophenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61694.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456706

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,5-Difluorophenyl)-3,4-difluorobenzenesulfonamide
305.25
MFCD15715989

Specifications

N-(3,5-Difluorophenyl)-3,4-difluorobenzenesulfonamide
97%
MFCD15715989
Solid
305.25
1g
Videos
SDS