missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3,5-Dichlorophenyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-(3,5-dichlorophenyl)benzenesulfonamide (C21) as an inhibitor of Rac activation by Dock5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-(3,5-dichlorophenyl)benzenesulfonamide (C21) as an inhibitor of Rac activation by Dock5.
Notes
Reacts with strong oxidizing agents. Store at room temperature.
N-(3,5-dichlorophenyl)benzenesulfonamide (C21) as an inhibitor of Rac activation by Dock5.
Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications
Color | White to Gray |
Quantity | 250 mg |
Formula Weight | 302.17 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | N-(3,5-Dichlorophenyl)benzenesulfonamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'