Alfa Aesar™ N-(3,5-Dichlorophenyl)-3,5-dinitrobenzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59531.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15435086

Quantity Price
1 £ 111.72 / 250mg
Estimated Shipment date
from Supplier 14-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(3,5-Dichlorophenyl)-3,5-dinitrobenzamide
Formula Weight 356.12
MDL Number MFCD00783875

Specification

Formula Weight 356.12
Quantity 250mg
Chemical Name or Material N-(3,5-Dichlorophenyl)-3,5-dinitrobenzamide
Assay 97%
Physical Form Solid
MDL Number MFCD00783875