missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,5-Dichlorophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61246.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497748

  • £109.18 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,5-Dichlorophenyl)-3,5-difluorobenzamide
302.11
MFCD04806518

Specifications

N-(3,5-Dichlorophenyl)-3,5-difluorobenzamide
97%
MFCD04806518
Solid
302.11
1g
Videos
SDS