missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[3,5-Bis(trifluoromethyl)phenyl]-4-bromo-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65347.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469576

  • £70.28 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-[3,5-Bis(trifluoromethyl)phenyl]-4-bromo-2-fluorobenzenesulfonamide
466.17
MFCD27975750

Specifications

N-[3,5-Bis(trifluoromethyl)phenyl]-4-bromo-2-fluorobenzenesulfonamide
97%
MFCD27975750
Solid
466.17
1g
Videos
SDS