missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dimethylphenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59918.03

Code : NEW

Additional Details : CAS Number : 36293-26-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426368

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dimethylphenyl)-3,5-dinitrobenzamide
315.28
2896129
36293-26-2
MFCD00791281

Specifications

N-(3,4-Dimethylphenyl)-3,5-dinitrobenzamide
Solid
315.28
2896129
1g
36293-26-2
97%
MFCD00791281
Off-white to cream
Videos
SDS