missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dimethylphenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62145.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15457086

  • £367.82 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dimethylphenyl)-3,5-difluorobenzenesulfonamide
297.32
MFCD26143310

Specifications

N-(3,4-Dimethylphenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD26143310
Solid
297.32
5g
Videos
SDS