missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dimethylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65598.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402579

  • £99.64 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dimethylphenyl)-2,2-diphenylacetamide
315.42
2141095
70298-69-0
MFCD00384380

Specifications

N-(3,4-Dimethylphenyl)-2,2-diphenylacetamide
Solid
315.42
2141095
70298-69-0
95%
MFCD00384380
1g
Videos
SDS