missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dimethoxyphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59617.03

Code : NEW

Additional Details : CAS Number : 449177-39-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15455998

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dimethoxyphenyl)-3-methylbenzamide
449177-39-3
271.32
MFCD00424036

Specifications

N-(3,4-Dimethoxyphenyl)-3-methylbenzamide
Solid
271.32
1g
449177-39-3
97%
MFCD00424036
Videos
SDS