missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Difluorophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59491.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485828

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Difluorophenyl)-3,5-dinitrobenzamide
323.21
MFCD01213248

Specifications

N-(3,4-Difluorophenyl)-3,5-dinitrobenzamide
97%
MFCD01213248
Solid
323.21
1g
Videos
SDS