missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dichlorophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59202.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485438

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dichlorophenyl)-3,5-dinitrobenzamide
356.12
MFCD00396208

Specifications

N-(3,4-Dichlorophenyl)-3,5-dinitrobenzamide
97%
MFCD00396208
Solid
356.12
1g
Videos
SDS