missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Dichlorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65666.03

Code : NEW

Additional Details : CAS Number : 50916-30-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432599

  • £119.78 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,4-Dichlorophenyl)-2,2-diphenylacetamide
356.25
2815957
50916-30-8
MFCD00028335

Specifications

N-(3,4-Dichlorophenyl)-2,2-diphenylacetamide
Solid
356.25
2815957
50916-30-8
95%
MFCD00028335
1g
Videos
SDS