Learn More
N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide, Tocris Bioscience™
σ1 selective ligand
Brand:  Tocris (Bio-Techne) 076310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13437973
  • £102.60 / 10mg
Estimated Shipment
09-07-2019
Description
Description
σ-1 selective ligand which binds with high affinity to human malignant melanoma cells A 2058.
Specifications
Chemical Identifiers
155054-42-5
358.223
4-iodo-n-piperidinoethylbenzamide, n-2-piperidinylamino ethyl-4-iodobenzamide, 4-iodo-n-2-1-piperidinyl ethyl benzamide, 4-iodo-n-2-piperidin-1-yl ethyl benzamide, n-2-piperidinylamino ethyl-4-iodobenzamide, 4-ipab, 125 i-pab, tocris-0763, i125-2-piperidinylaminoethyl-4-iodobenzamide, n-2-piperidinoethyl-4-iodobenzamide
4-iodo-N-(2-piperidin-1-ylethyl)benzamide
C14H19IN2O
WCMWVYQHPUQKHW-UHFFFAOYSA-N
127898
C1CCN(CC1)CCNC(=O)C2=CC=C(C=C2)I
Specifications
N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
155054-42-5
4-iodo-n-piperidinoethylbenzamide, n-2-piperidinylamino ethyl-4-iodobenzamide, 4-iodo-n-2-1-piperidinyl ethyl benzamide, 4-iodo-n-2-piperidin-1-yl ethyl benzamide, n-2-piperidinylamino ethyl-4-iodobenzamide, 4-ipab, 125 i-pab, tocris-0763, i125-2-piperidinylaminoethyl-4-iodobenzamide, n-2-piperidinoethyl-4-iodobenzamide
C1CCN(CC1)CCNC(=O)C2=CC=C(C=C2)I
358.223
358.22
10mg
C14H19IN2O
WCMWVYQHPUQKHW-UHFFFAOYSA-N
4-iodo-N-(2-piperidin-1-ylethyl)benzamide
127898
>98%
Videos