missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Methoxyphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60756.03

Code : NEW

Additional Details : CAS Number : 21226-32-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15467248

  • £358.49 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Methoxyphenyl)benzenesulfonamide
21226-32-4
263.31
MFCD00541868

Specifications

N-(2-Methoxyphenyl)benzenesulfonamide
1g
Solid
263.31
Off-white
21226-32-4
97%
MFCD00541868
Videos
SDS