missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Methoxyphenyl)-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60107.03

Code : NEW

Additional Details : CAS Number : 117561-97-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406588

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Methoxyphenyl)-2-nitrobenzenesulfonamide
308.31
MFCD00547739
3435947

Specifications

N-(2-Methoxyphenyl)-2-nitrobenzenesulfonamide
Solid
308.31
3435947
1g
97%
MFCD00547739
Videos
SDS