missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Methoxyphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65247.03

Code : NEW

Additional Details : CAS Number : 94869-90-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412499

  • £99.64 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Methoxyphenyl)-2,2-diphenylacetamide
317.39
2755953
94869-90-6
MFCD00387683

Specifications

N-(2-Methoxyphenyl)-2,2-diphenylacetamide
94869-90-6
95%
MFCD00387683
1g
Solid
317.39
2755953
Videos
SDS