missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65895.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489976

  • £91.90 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
293.41
MFCD28098837

Specifications

N-(2-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
293.41
1g
95%
MFCD28098837
Videos
SDS