missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Iodophenyl)-4-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59450.03

Code : NEW

Additional Details : CAS Number : 329939-01-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445778

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Iodophenyl)-4-methoxybenzamide
329939-01-7
353.16
MFCD00783646

Specifications

N-(2-Iodophenyl)-4-methoxybenzamide
1g
Solid
353.16
Light Sensitive
329939-01-7
97%
MFCD00783646
Videos
SDS