missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Iodophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59687.03

Code : NEW

Additional Details : CAS Number : 136138-26-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496068

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Iodophenyl)-2-methoxybenzamide
136138-26-6
353.16
MFCD00748349

Specifications

N-(2-Iodophenyl)-2-methoxybenzamide
1g
Solid
353.16
Light Sensitive
136138-26-6
97%
MFCD00748349
Videos
SDS