missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Iodophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65028.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442449

  • £91.90 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Iodophenyl)-2,2-diphenylacetamide
413.26
MFCD02662251

Specifications

N-(2-Iodophenyl)-2,2-diphenylacetamide
1g
95%
MFCD02662251
Light Sensitive
Solid
413.26
Videos
SDS