missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Hydroxyethyl)pyrrolidine, 97%, Thermo Scientific Chemicals
78.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Partly soluble in water. Soluble in alcohol, ether, benzene.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Partly soluble in water. Soluble in alcohol, ether, benzene.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Density | 0.985 |
Boiling Point | 79°C to 81°C |
Flash Point | 56°C (133°F) |
Refractive Index | 1.471 |
Quantity | 100 mL |
UN Number | UN1993 |
Beilstein | 102976 |
Solubility Information | Partly soluble in water. Soluble in alcohol,ether,benzene. |
Formula Weight | 115.17 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'