missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, 99%, Thermo Scientific Chemicals

Catalog Number 10455485
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3891-07-4
C10H9NO3
191.19
MFCD00005903
MWFLUYFYHANMCM-UHFFFAOYSA-N
2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
3354762
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
OCCN1C(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
3891-07-4
C10H9NO3
191.19
MFCD00005903
MWFLUYFYHANMCM-UHFFFAOYSA-N
2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
3354762
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
OCCN1C(=O)C2=CC=CC=C2C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 126°C to 129°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (HPLC)
Packaging Glass bottle
Quantity 500 g
Formula Weight 191.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'