missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)morpholine, 99%, ACROS Organics™

£11.10 - £31.10

Chemical Identifiers

CAS 622-40-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.17
MDL Number MFCD00006180
InChI Key KKFDCBRMNNSAAW-UHFFFAOYSA-N
Synonym 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
PubChem CID 61163
ChEBI CHEBI:67144
IUPAC Name 2-morpholin-4-ylethanol
SMILES C1COCCN1CCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10598470
SDS View Documents
Acros Organics
121180050
5g Glass bottle
£11.10
5g
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2021
Please to purchase this item. Need a web account? Register with us today!
10041070
SDS View Documents
Acros Organics
121181000
100g Glass bottle
£13.20
100g
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2021
Please to purchase this item. Need a web account? Register with us today!
10605742
SDS View Documents
Acros Organics
121185000
500g Glass bottle
£31.10
500g
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

622-40-2
131.17
KKFDCBRMNNSAAW-UHFFFAOYSA-N
61163
2-morpholin-4-ylethanol
C6H13NO2
MFCD00006180
2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
CHEBI:67144
C1COCCN1CCO

Specifications

N-(2-Hydroxyethyl)morpholine
107°C
Glass bottle
1.08
0.5% max.
Colorless to Yellow
5g
622-40-2
100.0
C6H13NO2
27, 7
Solubility in water: miscible
C1COCCN1CCO
131.17
CHEBI:67144
Liquid
1.0800g/mL
Authentic
1.4760 to 1.4780
26.6 mPa.s (20°C)
223.0°C to 225.0°C
-1.0°C
99%
98.5
99%
MFCD00006180
2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
KKFDCBRMNNSAAW-UHFFFAOYSA-N
2-morpholin-4-ylethanol
61163
131.17
99%
Safety and Handling

Safety and Handling

Product Identifier
  • N-(2-Hydroxyethyl)morpholine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications