missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Fluorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65151.03

Code : NEW

Additional Details : CAS Number : 198337-86-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402479

  • £91.90 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Fluorophenyl)-2,2-diphenylacetamide
305.35
22878703
198337-86-9
MFCD00090667

Specifications

N-(2-Fluorophenyl)-2,2-diphenylacetamide
Solid
305.35
22878703
198337-86-9
95%
MFCD00090667
1g
Videos
SDS