Alfa Aesar™ N-(2-Fluoro-4-iodophenyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H60728.MD

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15435976

Quantity Price
1 £ 111.72 / 250mg
Estimated Shipment date
from Supplier 14-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(2-Fluoro-4-iodophenyl)benzenesulfonamide
Formula Weight 377.17
MDL Number MFCD22683146

Specification

Formula Weight 377.17
MDL Number MFCD22683146
Physical Form Solid
Assay 97%
Sensitivity Light Sensitive
Chemical Name or Material N-(2-Fluoro-4-iodophenyl)benzenesulfonamide
Quantity 250mg