Alfa Aesar™ N-(2-Fluoro-4-iodophenyl)-2,2-diphenylacetamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65011.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15449316

Quantity Price
1 £ 23.0 / 250mg
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(2-Fluoro-4-iodophenyl)-2,2-diphenylacetamide
Formula Weight 431.25
MDL Number MFCD05851457

Specification

Formula Weight 431.25
Sensitivity Light Sensitive
MDL Number MFCD05851457
Chemical Name or Material N-(2-Fluoro-4-iodophenyl)-2,2-diphenylacetamide
Quantity 250mg
Physical Form Solid
Assay 95%
Documentation

Technical documents