missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Fluoro-4-chlorophenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59720.03

Code : NEW

Additional Details : CAS Number : 546067-98-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446118

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Fluoro-4-chlorophenyl)-3-methylbenzamide
546067-98-5
263.7
MFCD03392612

Specifications

N-(2-Fluoro-4-chlorophenyl)-3-methylbenzamide
Solid
263.7
1g
546067-98-5
97%
MFCD03392612
Videos
SDS