missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Fluoro-3-methylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65136.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409406

  • £91.90 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Fluoro-3-methylphenyl)-2,2-diphenylacetamide
319.38
MFCD28098753

Specifications

N-(2-Fluoro-3-methylphenyl)-2,2-diphenylacetamide
95%
MFCD28098753
Solid
319.38
1g
Videos
SDS