missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Ethylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65766.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489886

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Ethylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
279.38
MFCD28098822

Specifications

N-(2-Ethylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
MFCD28098822
Solid
279.38
1g
Videos
SDS