missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Ethylphenyl)-4-nitrobenzamide, 97%

Brand:  Alfa Aesar™ H60214.03

Code : NEW

Additional Details : CAS Number : 35709-77-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406698

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Ethylphenyl)-4-nitrobenzamide
C15H14N2O3
MFCD00171511
35709-77-4
270.28

Specifications

N-(2-Ethylphenyl)-4-nitrobenzamide
97%
270.28
1g
35709-77-4
C15H14N2O3
MFCD00171511
Videos
SDS