missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Ethylphenyl)-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60552.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497028

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Ethylphenyl)-2-nitrobenzenesulfonamide
306.33
MFCD01211980

Specifications

N-(2-Ethylphenyl)-2-nitrobenzenesulfonamide
97%
MFCD01211980
1g
Solid
306.33
Red
Videos
SDS