missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Ethylhexyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65341.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412529

  • £119.78 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Ethylhexyl)-2,2-diphenylacetamide
323.48
MFCD01349496

Specifications

N-(2-Ethylhexyl)-2,2-diphenylacetamide
95%
MFCD01349496
Solid
323.48
1g
Videos
SDS