missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Ethoxyphenyl)-4-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59805.03

Code : NEW

Additional Details : CAS Number : 303795-58-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476218

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Ethoxyphenyl)-4-methoxybenzamide
303795-58-6
271.32
MFCD00522613

Specifications

N-(2-Ethoxyphenyl)-4-methoxybenzamide
Solid
271.32
1g
303795-58-6
97%
MFCD00522613
Videos
SDS