missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chlorophenyl)maleimide, 97%, Thermo Scientific™

Catalog Number 15575236
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1203-24-3
C10H6ClNO2
207.62
KPQOXMCRYWDRSB-UHFFFAOYSA-N
1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide
14550
1-(2-chlorophenyl)pyrrole-2,5-dione
C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl
This item is not returnable. View return policy
1203-24-3
C10H6ClNO2
207.62
KPQOXMCRYWDRSB-UHFFFAOYSA-N
1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide
14550
1-(2-chlorophenyl)pyrrole-2,5-dione
C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl
Melting Point 108.0°C to 109.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 207.62
Percent Purity 97%
Chemical Name or Material N-(2-Chlorophenyl)maleimide
compliance-icons
Product Identifier
  • N-(2-Chlorophenyl)maleimide, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'