missing translation for 'onlineSavingsMsg'
Learn More

N-(2-chloroethyl)dibenzylamine hydrochloride, 98%, Thermo Scientific™

Catalog Number p-4228502
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
55-43-6
C16H19Cl2N
296.24
MFCD00012518
LZXCEBPGNFLHEQ-UHFFFAOYSA-N
dibenamine hydrochloride, n-2-chloroethyl dibenzylamine hydrochloride, sympatholytin, n,n-dibenzyl-2-chloroethanamine hydrochloride, n,n-dibenzyl-2-chloroethylamine hydrochloride, dibenzylchlorethamine hydrochloride, dibenzylchlorethylamine hydrochloride, n,n-dibenzylaminoethyl chloride hydrochloride, 2-chloroethyldibenzylamine hydrochloride, 2-dibenzylaminoethyl chloride hydrochloride
5925
[H+].[Cl-].ClCCN(CC1=CC=CC=C1)CC1=CC=CC=C1
This item is not returnable. View return policy
55-43-6
C16H19Cl2N
296.24
MFCD00012518
LZXCEBPGNFLHEQ-UHFFFAOYSA-N
dibenamine hydrochloride, n-2-chloroethyl dibenzylamine hydrochloride, sympatholytin, n,n-dibenzyl-2-chloroethanamine hydrochloride, n,n-dibenzyl-2-chloroethylamine hydrochloride, dibenzylchlorethamine hydrochloride, dibenzylchlorethylamine hydrochloride, n,n-dibenzylaminoethyl chloride hydrochloride, 2-chloroethyldibenzylamine hydrochloride, 2-dibenzylaminoethyl chloride hydrochloride
5925
[H+].[Cl-].ClCCN(CC1=CC=CC=C1)CC1=CC=CC=C1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 188°C to 194°C
Color White
Elemental Analysis 63.2 to 66.5% C, 4.5 to 4.9% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3009
IUPAC Name hydrogen dibenzyl(2-chloroethyl)amine chloride
Formula Weight 296.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-chloroethyl)dibenzylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Chloroethyl)dibenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'