Alfa Aesar™ N-[2-Chloro-4-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H61252.MD

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15435515

Quantity Price
1 £ 29.89 / 250mg
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-[2-Chloro-4-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide
Formula Weight 371.71

Specification

Formula Weight 371.71
Quantity 250mg
Physical Form Solid
Assay 97%
Chemical Name or Material N-[2-Chloro-4-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide
Documentation

Technical documents